Learn More
Alkali Scientific DTT [DL Dithiothreitol] (Cleland's Reagent)], 25 G
SDP
Supplier:  Alkali Scientific DTT004CP
Encompass_Preferred
Dithiothreitol (DTT), also known as Cleland's Reagent, is a reducing agent widely used to stabilize proteins and enzymes in laboratory research. This 25g reagent is crucial for preventing oxidative damage in protein samples by reducing disulfide bonds, allowing proteins to retain their functional, reduced states. DTT is essential in protein purification, structural analysis, and enzymatic assays. It is frequently used in molecular biology applications, including protein extraction, electrophoresis, and chromatography, to ensure that proteins remain active and undamaged by oxidation during experimental procedures. Researchers rely on DTT for accurate and reproducible results in protein analysis and enzyme activity studies.
Catalog No. 50-006-60990