Learn More
Sino Biological Assay Buffer and Co-factors: DTT Solution
Catalog No. 502569385
Encompass_Preferred
0.1 M DTT solution used for preparing protein kinase assay buffer.
Catalog No. 50-256-9385 Supplier Sino Biological Supplier No. D8609B10
0.1 M DTT solution used for preparing protein kinase assay buffer.