Learn More
Sino Biological Assay Buffer and Co-factors: DTT Solution
Catalog No. 502569386
Encompass_Preferred
1 M DTT solution used for preparing protein kinase assay buffer.
Catalog No. 50-256-9386 Supplier Sino Biological Supplier No. D860910
1 M DTT solution used for preparing protein kinase assay buffer.