Learn More
TRIBIOSCIENCE INC DTT Solution (1M), DNase / RNase Free, 50 ML
SDP
Catalog No. NC3454597
Encompass_Preferred
Tribioscience’s Dithiothreitol (DTT) is used in proteomics applications to maintain sulfhydryl (-SH) groups in the reduced state and for quantitative reduction of disulfide (-S-S-) groups. By reducing the disulfide bonds in a protein sample, the protein can be more effectively fragmented and analyzed. The concentration is 1M.
Catalog No. NC3454597 Supplier TRIBIOSCIENCE INC Supplier No. TBS5039
Tribioscience’s Dithiothreitol (DTT) is used in proteomics applications to maintain sulfhydryl (-SH) groups in the reduced state and for quantitative reduction of disulfide (-S-S-) groups. By reducing the disulfide bonds in a protein sample, the protein can be more effectively fragmented and analyzed. The concentration is 1M.