Learn More
Epigentek Inc DTT Solution (1 ml, 500 mM)
SDP
Catalog No. 501097107
Encompass_Preferred
DTT, also known as Cleland's Reagent, is a particularly strong reducing agent, owing to its high conformational proprensity to form a six member ring with an internal disulfide bond. It has a redox potential of -0.33 V @ pH 7. It is used for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE). The 500 mM stock solution can be directly used to achieve the amount of reducing agent required for your specific application.
Catalog No. 50-109-7107 Supplier Epigentek Inc Supplier No. R11041
DTT, also known as Cleland's Reagent, is a particularly strong reducing agent, owing to its high conformational proprensity to form a six member ring with an internal disulfide bond. It has a redox potential of -0.33 V @ pH 7. It is used for many protein research applications, including reducing polyacrylamide electrophoresis (SDS-PAGE). The 500 mM stock solution can be directly used to achieve the amount of reducing agent required for your specific application.