Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DTT / TG11 [Dithiothreitol + alpha-Thioglycerol] (1:1 mixture) [Matrix for FABMS and liquid SIMS], TCI America™
SDP

Catalog No. S04361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
S04361G 1 g
1 options

Catalog No. S04361G

Supplier: TCI America S04361G

Chemical Identifiers

MDL Number MFCD01321186
Synonym Dithiothreitol + alpha-Thioglycerol

Specifications

Quantity 1 g
Chemical Name or Material DTT / TG11 [Dithiothreitol + alpha-Thioglycerol] (1:1 mixture) [Matrix for FABMS and liquid SIM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.