Learn More
AdipoGen DTTCI
SDP
Supplier:  AdipoGen CDXD0253G001
Encompass_Preferred
Chemical. CAS 3071-70-3. Formula C25H25N2S2I. MW 544.52. Synthetic. Near-IR NIR dye with maximal absorbance at 772nm.
Catalog No. 50-256-1166