Learn More
American Radiolabeled Chemicals Inc DTTP ME-3H A 5LC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1881A5MC
Encompass
Deoxythymidine 5'-methyl-3H triphosphate tetrasodium salt (dTTP) 5mCi, 70-90 Ci/mmol, ethanol:water (1:1), 1 mCi/ml, M.W. 570.6, Shipped in Dry Ice
Catalog No. 50-753-2694
Add to Cart
Request a Quote