Learn More
Revvity Health Sciences Inc dTTP, [α-32P]- 800Ci/mmol 10mCi/ml EasyTide, 250 µCi
Catalog No. 502810311
Encompass
dTTP, [α-32P]- 800Ci/mmol 10mCi/ml EasyTide, 250 µCi
Catalog No. 50-281-0311 Supplier Revvity Health Sciences Inc Supplier No. BLU505A250UC
dTTP, [α-32P]- 800Ci/mmol 10mCi/ml EasyTide, 250 µCi