Learn More
Aviva Systems Biology Corp DTX1 Antibody - N-terminal region (ARP38458_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38458_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DTX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DTX1. Alternate names: hDx-1ApplicationWB. Species reactivity: Cow, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56663