Learn More
Aviva Systems Biology Corp Dtx3 Antibody - C-terminal region (ARP43465_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43465_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dtx3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: Deltex3, MGC106339, mg310ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-285-075