Learn More
Electron Microscopy Sciences Microwave Transparent Dummy Load Container
SDP
Catalog No. 5030577
Encompass
Microwave transparent "dummy load" vented containers are ideal for use in light or electron microscopy microwave laboratory procedures.
Catalog No. 50-305-77 Supplier Electron Microscopy Sciences Supplier No. 97088
Add to Cart
Add to Cart
Microwave transparent "dummy load" vented containers are ideal for use in light or electron microscopy microwave laboratory procedures.