Learn More
MSC Milwaukee Tool Wet/Dry Vacuum, 9 gal., Dual-Battery, 1 ea
Catalog No. 502866835
Encompass
This wet/dry vacuum is designed for efficient cleanup in various environments. It features a dual-battery system for extended operation and a durable plastic tank for robust performance.
  • Battery powered
  • Standard filtration type
  • Collection capacity of 9.0 gal
  • Bagless vacuum collection
  • Plastic tank material
  • Maximum air flow of 109 CFM
  • Hose diameter of 1.8 in
  • Horse power of 4.2
  • Includes motor head, tank, cart, high efficiency filter, flexible hose, crevice tool, utility nozzle, and extension wands
Catalog No. 50-286-6835 Supplier MSC Supplier No. 20945994
This wet/dry vacuum is designed for efficient cleanup in various environments. It features a dual-battery system for extended operation and a durable plastic tank for robust performance.
  • Battery powered
  • Standard filtration type
  • Collection capacity of 9.0 gal
  • Bagless vacuum collection
  • Plastic tank material
  • Maximum air flow of 109 CFM
  • Hose diameter of 1.8 in
  • Horse power of 4.2
  • Includes motor head, tank, cart, high efficiency filter, flexible hose, crevice tool, utility nozzle, and extension wands