Learn More
Bellco Glass, Inc. Dual CellTrol,pH/DO/Tem/SP 4 Pumps
SDP
Catalog No. 501946362
Encompass_Preferred
The CellTrol II control module monitors bioreactors process values and performs PLC (Allen-Bradley Programmable Logic Controller) based process control. A touch screen operator interface provides access to all PLC based control functions andallows viewing and modification of control parameters. Signals types include temperature, pH, DO, speed, and foam/level. PLC control drives the inc/dec outlets assigned to each channel as well as to pumps mounted on the front of the unit. The pump assignments are user defined from the operator interface. Cell-Soft Software are also available. Optional tethered speed drive, optional single support flow station and weight platforms are sold separately.. MADE IN THE USA
Catalog No. 50-194-6362 Supplier Bellco Glass, Inc. Supplier No. 780520400
Add to Cart
Add to Cart
Request a Quote
The CellTrol II control module monitors bioreactors process values and performs PLC (Allen-Bradley Programmable Logic Controller) based process control. A touch screen operator interface provides access to all PLC based control functions andallows viewing and modification of control parameters. Signals types include temperature, pH, DO, speed, and foam/level. PLC control drives the inc/dec outlets assigned to each channel as well as to pumps mounted on the front of the unit. The pump assignments are user defined from the operator interface. Cell-Soft Software are also available. Optional tethered speed drive, optional single support flow station and weight platforms are sold separately.. MADE IN THE USA