Learn More
Cole-Parmer Revolutionary Science Dual-Chamber Poly Water Bath, 5.5L (each), 90 C; 120VAC
Catalog No. NC1405418 Shop All Cole-Parmer Products
Encompass
This economical dual-chamber bath provides you the flexibility of performing two different temperature based experiments simultaneously. Separate chambers allow you to set the temperature independently from ambient to 90° C, without temperature interference from the other bath. The dual 5.5 liter chambers allow you to heat samples of varying sizes. Each chamber has its own digital temperature display for easy setting and read-out. Each chamber interior is made from chemical and corrosion-resistant polypropylene, allowing for easy clean-up and durability. The rugged exterior resists dents, cracking, and chemicals. Low-level sensor automatically shuts the bath down and sounds an alarm if the water level goes below 1-inch.
Catalog No. NC1405418 Supplier Cole-Parmer Supplier No. 3906650
This economical dual-chamber bath provides you the flexibility of performing two different temperature based experiments simultaneously. Separate chambers allow you to set the temperature independently from ambient to 90° C, without temperature interference from the other bath. The dual 5.5 liter chambers allow you to heat samples of varying sizes. Each chamber has its own digital temperature display for easy setting and read-out. Each chamber interior is made from chemical and corrosion-resistant polypropylene, allowing for easy clean-up and durability. The rugged exterior resists dents, cracking, and chemicals. Low-level sensor automatically shuts the bath down and sounds an alarm if the water level goes below 1-inch.