Learn More
Chemglass Life Sciences Vacuum Trap, Dual Chamber, Complete
SDP
Catalog No. 501531744
Encompass_Preferred
Vacuum Trap, Complete, 60mm Body OD x 300mm Overall Length, 30mm OD Side and Inner Tubes, 55/50 Joint. Similar to CG-4514, but with the top inlet tube bent at a 90 angle.
Catalog No. 50-153-1744 Supplier Chemglass Life Sciences Supplier No. CG452501
Vacuum Trap, Complete, 60mm Body OD x 300mm Overall Length, 30mm OD Side and Inner Tubes, 55/50 Joint. Similar to CG-4514, but with the top inlet tube bent at a 90 angle.