Learn More
Braintree Scientific Dual Digital Lab Std Stereotaxic w/Mouse and Rat Adaptors
SDP
Catalog No. 501953998
Encompass
Digital Lab Standard™ features a sealed electronic sensor, highly immune to interference and environmental conditions. Movements are monitored by a large, easy-to-read LED display module and are accurate to 10uL. A zeroing function allows the user to target specific coordinates in the brain for injection, implantation, etc. Calculation of distance measurements via the vernier scales is thus no longer necessary...saving the user valuable time and improving accuracy. **Sold for research purposes only and not for human use**
Catalog No. 50-195-3998 Supplier Braintree Scientific Supplier No. 51970
Add to Cart
Add to Cart
Request a Quote
Digital Lab Standard™ features a sealed electronic sensor, highly immune to interference and environmental conditions. Movements are monitored by a large, easy-to-read LED display module and are accurate to 10uL. A zeroing function allows the user to target specific coordinates in the brain for injection, implantation, etc. Calculation of distance measurements via the vernier scales is thus no longer necessary...saving the user valuable time and improving accuracy. **Sold for research purposes only and not for human use**