Learn More
TARGETMOL CHEMICALS INC DUAL FAAH/SEH-IN-1 5MG
SDP
Catalog No. 502579418
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dual FAAH/sEH-IN-1 is a dual inhibitor of sEH (soluble epoxide hydrolase) (IC50 9.6 nM) and FAAH (fatty acid amide hydrolase) (IC50 7 nM) with high affinity and anti-inflammatory activity. purity: 99%
Catalog No. 50-257-9418 Supplier TARGETMOL CHEMICALS INC Supplier No. T635365MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dual FAAH/sEH-IN-1 is a dual inhibitor of sEH (soluble epoxide hydrolase) (IC50 9.6 nM) and FAAH (fatty acid amide hydrolase) (IC50 7 nM) with high affinity and anti-inflammatory activity. purity: 99%