Learn More
MSC Buyers Products Dual Flow Hydraulic Spreader Valve, 3/4 Inch NPT, 7-15 GPM
Supplier: MSC 45780962
This dual flow hydraulic spreader valve offers efficient and precise flow control. Its compact, rugged aluminum body allows for easy installation with minimal plumbing. It integrates a differential style relief valve, on/off control, and pressure compensation for reliable performance. Flow is easily adjusted via detent hand knobs with 11 positive positions. All primary ports are 3/4 inch NPT, handling 7-15 GPM flow with a 2000 PSI max pressure and 1500 PSI relief setting.
- Compact, easy installation with minimal plumbing.
- Rugged, weather-resistant aluminum body.
- Built-in system relief and on/off control.
- Pressure compensated for consistent flow.
- Two detent knobs, 11 positive flow adjustment positions.
- Primary ports are 3/4 inch NPT.
- Relief cartridge standard setting: 1500 PSI (105 bar).
- Flow rate: 7-15 GPM.
- Maximum operating pressure: 2000 PSI (140 bar).