Learn More
Grainger Dual Foot Air Chuck 14 Long
Catalog No. 502750886
Encompass_Preferred
The Haltec Dual Foot Air Chuck is a pneumatic tool accessory used to inflate tires. It has a 1/4 inch NPT inlet and two flexible rubber hoses with chucks to connect to tire valves. The chucks have pressure bleeder valves and swivel 360 degrees. The unit is made of zinc plated steel and has a 150 PSI maximum working pressure. It can inflate two tires at once and the hoses are 12 inches long. This air chuck allows hands-free tire inflation.
Catalog No. 50-275-0886 Supplier Grainger Supplier No. 140X18
The Haltec Dual Foot Air Chuck is a pneumatic tool accessory used to inflate tires. It has a 1/4 inch NPT inlet and two flexible rubber hoses with chucks to connect to tire valves. The chucks have pressure bleeder valves and swivel 360 degrees. The unit is made of zinc plated steel and has a 150 PSI maximum working pressure. It can inflate two tires at once and the hoses are 12 inches long. This air chuck allows hands-free tire inflation.