Learn More
Grainger Dual Foot Air Chuck w/Thread Nut
Catalog No. 502750769
Encompass_Preferred
The Haltec Dual Foot Air Chuck is a pneumatic tool accessory used to connect air hoses to pneumatic tools that require a 1/4" NPT inlet. It has a zinc die-cast body with rubber feet to prevent sliding. The chuck has a 1/4" NPT female inlet and accepts hoses with a 1/4" ID. It can operate at pressures up to 300 PSI. This air chuck provides a secure connection for air supply hoses. Its dual foot design holds hoses in place during use.
Catalog No. 50-275-0769 Supplier Grainger Supplier No. 140X04
The Haltec Dual Foot Air Chuck is a pneumatic tool accessory used to connect air hoses to pneumatic tools that require a 1/4" NPT inlet. It has a zinc die-cast body with rubber feet to prevent sliding. The chuck has a 1/4" NPT female inlet and accepts hoses with a 1/4" ID. It can operate at pressures up to 300 PSI. This air chuck provides a secure connection for air supply hoses. Its dual foot design holds hoses in place during use.