Learn More
Stoelting™ Dual Gooseneck Illuminator
SDP
Catalog No. 10001050 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Dual gooseneck
Catalog No. Model
10-001-050 Dual gooseneck
1 options
Catalog No. 10-001-050 Supplier Stoelting™ Supplier No. 59259
Add to Cart
Add to Cart
Dual Gooseneck Illuminator is a rugged, convection-cooled enclosure that results in no noise or vibration.
  • Units deliver over 10,000 foot candles of intense, cold illumination
  • Easy lamp changes and a lifetime warranty round out these hardworking illumination
  • Dual-Pipe Fiber Optic allows the researcher to supply light from both sides, to eliminate shadows and mounts directly without adaptors.
Specifications
For Use With (Application) Neuroscience
Voltage 150 w
Model Dual gooseneck