Learn More
GUARDIAN EQUIPMENT Eye Wash/Drench Hose Unit, Deck Mounted
Catalog No. NC0393200
Encompass_Preferred
Dual purpose eyewash/drench hose for deck mounting. Unit meets the provisions of ANSI Z358.1-2014 as both an eyewash and a drench hose. Unit may be left in the deck flange for use as a fixed eyewash, leaving user’s hands free. Alternatively, unit may be removed for use as a drench hose to rinse any part of user’s eyes, face or body.
Catalog No. NC0393200 Supplier GUARDIAN EQUIPMENT Supplier No. G5022
Dual purpose eyewash/drench hose for deck mounting. Unit meets the provisions of ANSI Z358.1-2014 as both an eyewash and a drench hose. Unit may be left in the deck flange for use as a fixed eyewash, leaving user’s hands free. Alternatively, unit may be removed for use as a drench hose to rinse any part of user’s eyes, face or body.