Learn More
Thermo Scientific™ Universal Clamps for MaxQ 4000, 4450, 6000, and Solaris Shakers
Catalog No. 01258057 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Catalog No. 01-258-057 Supplier Thermo Scientific™ Supplier No. 75004102
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mix and match these shaker platform clamps, mats and racks, available in a variety of sizes, to suit individual needs in the laboratory.
Mix and match Thermo Scientific™ MaxQ™ Shaker Universal Clamps, available in a variety of sizes, to suit individual needs in the laboratory. Compatible with MaxQ shaker platforms.
Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory.
TRUSTED_SUSTAINABILITY
Specifications
Holds Vessel
Product Type Dual Height Adjustable Clamp
Dimensions (L x W) 387 x 326 mm
Dimensions (LxW) Platform 12 x 14 in.
For Use With (Equipment) Solaris Open Air and Temperature Controlled Shakers, MaxQ6000 Shakers