Learn More
Enzo Life Sciences Dual-iODN (class I) (endotoxin-free) (synthetic) (BULK) (1mg)
SDP
Supplier:  Enzo Life Sciences ALX746355M001
Encompass_Preferred
Inhibitory iODN oligodeoxynucleotide with phosphorothioate backbone. Sequence of an inhibitory ODN for in vivo use in rodents (50-150µg per injection): prototype class I that potently inhibits TLR9 and TLR7 signaling. Formulation: Lyophilized. Sterile. Long Term Storage: +4°C.
Catalog No. 50-200-9679