Learn More
World Precision Instrument Dual Large Probe Holder
SDP
Catalog No. 502539158
Encompass
  • The Double-Head Large Probe Holder has a large ”C” gripping component at each end, with the “V” clamp attachment.
  • Dual heads allow a secure hold for two different components. Clamps ¼"–½" diameter.
Catalog No. 50-253-9158 Supplier World Precision Instrument Supplier No. 505257
  • The Double-Head Large Probe Holder has a large ”C” gripping component at each end, with the “V” clamp attachment.
  • Dual heads allow a secure hold for two different components. Clamps ¼"–½" diameter.