Learn More
Stoelting™ Dual Large Probe Holder
SDP
Catalog No. 10000163 Shop All Stoelting Co Products
Change view
Click to view available options
For Use With (Application):
Neuroscience
Catalog No. For Use With (Application)
10-000-163 Neuroscience
1 options
Catalog No. 10-000-163 Supplier Stoelting™ Supplier No. 51638
 Restricted Item
Add to Cart
Add to Cart
Stoelting Dual Large Probe Holder has a large C gripping component at each end, with the V clamp attachment.
  • Dual heads allow a secure hold for two different components.
  • Clamps 0.25 to 0.5 inch diameter
Specifications
For Use With (Application) Neuroscience