Learn More
Grainger Dual Led Light w/Magnetic Brackets 24
Catalog No. 502753115
Encompass_Preferred
The Grainger 157G35 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and sensitivity, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery life of up to 3000 hours. The Speedglas Welding Helmet is designed for reliability and durability on the job.
Catalog No. 50-275-3115 Supplier Grainger Supplier No. 157G35
The Grainger 157G35 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face during welding, cutting and grinding. It has a large viewing area, multiple adjustable settings like shade control and sensitivity, and comfortable padded headgear. The helmet provides shade levels from 5 to 13 for optimal eye protection. It meets industry safety standards and has a battery life of up to 3000 hours. The Speedglas Welding Helmet is designed for reliability and durability on the job.