Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dual Lens Magnifiers
SCI_ED

Catalog No. S19230B
Change view
Click to view available options
Magnification:
3x, 6x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S19230B 3x, 6x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S19230B Supplier Learning Resources, Inc Supplier No. LER2046
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ten versatile plastic magnifiers.

  • Feature 3X and 6X magnifications
  • Measure: 1.75 in. (4.4cm) diameter
  • Grades K+

Order Info

Ten per pack

Specifications

Magnification 3x, 6x
Diameter (Metric) Outer 4.44 cm
Product Type Magnifier, Dual Lens
Material Plastic
Diameter (English) Outer 1.75 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.