Learn More
TA Instruments Dual Liquid Light Guide, 1 Meter Length, 5 mm Diameter, for PCA Accessory, for ARES Rheometer, Temperature Range-50 to + 80 Degrees Celsius, Requires RCS Cooling Accessory for Sub-ambient Operation
Catalog No. 5000580600 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Dual Liquid Light Guide is a 1 meter long, 5 mm diameter dual liquid light guide for the PCA Accessory, for the ARES rheometer. The temperature range of use is -50 to +80 degrees Celsius. For sub-ambient temperature operation, the RCS Cooling Accessory is required.
Catalog No. 50-005-80600 Supplier TA Instruments Supplier No. 935012.001
Waters TA Instruments Dual Liquid Light Guide is a 1 meter long, 5 mm diameter dual liquid light guide for the PCA Accessory, for the ARES rheometer. The temperature range of use is -50 to +80 degrees Celsius. For sub-ambient temperature operation, the RCS Cooling Accessory is required.