Learn More
BPS Bioscience Inc Dual Luciferase Assay System, 100 ml, 1000 assays (at 100 μl), Firefly and Renilla luciferase targets, high-throughput, luminescence detection
SDP
Catalog No. NC1748022
Encompass
Dual Luciferase Assay System, 100 ml, 1000 assays (at 100 μl), Firefly and Renilla luciferase targets, high-throughput, luminescence detection
Catalog No. NC1748022 Supplier BPS Bioscience Inc Supplier No. 606832
Dual Luciferase Assay System, 100 ml, 1000 assays (at 100 μl), Firefly and Renilla luciferase targets, high-throughput, luminescence detection