Learn More
BPS Bioscience Inc DUAL LUCIFERASE FIREFLY-RENIL
SDP
Catalog No. NC1805677
Encompass
10ml -Dual Luciferase (Firefly-Renilla) Assay System. used for high-throughput, rapid quantitation of both Firefly and Renilla luciferases from a single sample in mammalian cell culture.
Catalog No. NC1805677 Supplier BPS Bioscience Inc Supplier No. 606831
10ml -Dual Luciferase (Firefly-Renilla) Assay System. used for high-throughput, rapid quantitation of both Firefly and Renilla luciferases from a single sample in mammalian cell culture.