Learn More
National Optical Dual Magnification (1X/3X) Stereo Microscope
SCI_ED
Catalog No. S81782
Change view
Click to view available options
Magnification Power:
1X, 3X
Catalog No. Magnification Power
S81782 1X, 3X
1 options
Catalog No. S81782 Supplier National Optical Supplier No. 446TBL10
Add to Cart
Add to Cart
All the quality features and versatility of larger stereo microscopes, in a compact size that is more efficient for many uses. Choose low or high magnification simply by turning the objective turret.
All the quality features and versatility of larger stereo microscopes, in a compact size that is more efficient for many uses. Choose low or high magnification simply by turning the objective turret.
MAGNIFICATION
  • 1x & 3x
OPTICAL SYSTEM
  • Paired widefield eyepieces with diopter adjustment on left side. Interpupillary adjustment from 55 mm to 75 mm. Reticle holder/retainer accepts eyepiece reticle with 23 mm O.D.
  • Magnification changed by rotating objective turret 90 degrees, to positive click stop.
  • Working distance: 47 mm. Maximum specimen height on stage, 35 mm
  • Head rotates 360 degrees to permit reverse head position viewing.
ILLUMINATION
  • High intensity 12 V, 10 W incidental and transmitted illumination is built-in.
  • Separate base mounted switches control power on/off, top and bottom lights, permit use of top light only, bottom light only, top and bottom lights together.
FOCUSING
  • Rack & pinion focusing has slip clutch to protect mechanism against over-turning, and special tension adjustment.
STAGE
  • Supplied with two 80 mm stage plates, one frosted glass and one plastic black/white reversible contrast. Two locked-on stage clips.
Specifications
Voltage 110/220 V
Magnification Power 1X, 3X
Objective Types Dual
Product Type Stereo Microscope
Model 446TBL-10
Head Style Binocular
Lighting Type Halogen Upper and Lower
Eyepiece Paired WF 10X
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov