Learn More
National Optical Dual Magnification Digital Stereo with Built-in 3.0 MP Camera
SCI_ED
Catalog No. S81784 Shop All Motic Swift Line Products
Change view
Click to view available options
Magnification Power:
1X, 3X
Catalog No. Magnification Power
S81784 1X, 3X
1 options
Catalog No. S81784 Supplier National Optical Supplier No. DC4456H
Add to Cart
Add to Cart
A very versatile stereoscope microscope that permits you to change from a widefield image at 10X magnification to the close-up detail of 30X magnification simply by rotating the objective turret. Built in high-resolution digital camera offers 3.0 MP live image resolution at 1280 x 1024.
A very versatile stereoscope microscope that permits you to change from a widefield image at 10x magnification to the close-up detail of 30x magnification simply by rotating the objective turret. The extra rugged fixed-arm stand has both incidental and transmitted illumination. Built in high-resolution digital camera offers 3.0 megapixel live image resolution at 1280 x 1024.  Supplied with Motic Images software, which allows the user to view, capture, label, manipulate and measure images.  The microscope is fully functional, independent of the camera application.
OPTICAL SYSTEM
  • Paired widefield eyepieces with diopter adjustment on left side. Interpupillary adjustment from 55 mm to 75 mm. Reticle holder/retainer accepts eyepiece reticle with 23 mm O.D.
  • Paired objectives are parfocalled, parcentered, achromatic.
  • Magnification changed by rotating objective turret 90 degrees.
  • Working distance: 75 mm. Maximum specimen height: 35 mm.
ILLUMINATION
  • High intensity, built-in 12 V 10 W halogen, incidental illumination and 12 V 10 W tungsten, transmitted illumination.
  • 12 V DC power converter operates on 100 V to 240 V 50/60 Hz current, without need of additional transformer.
  • Separate base mounted switches control power on/off, top and bottom lights.  This permits use of top light only, bottom light only, or top and bottom lights together.
FOCUSING
  • Rack and pinion focusing has slip clutch to protect mechanism against over-turning, and special tension adjustment.
STAGE
  • Supplied with two 80 mm stage plates, one frosted glass plate and one plastic black/white reversible contrast plate.
  • Two locked-on stage clips.
Specifications
Voltage 110/220 V
Magnification Power 1X, 3X
Objective Types Dual
Product Type Digital Stereo Microscope
Model DC4-456 H
Head Style Binocular Digital
Lighting Type Halogen Upper, Fluorescent Lower
Eyepiece Paired WF 10X
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov