Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motic™ Swift Line Dual Magnification Intermediate Stereo Microscopes
SCI_ED

Catalog No. p-7184534 Shop All Motic Swift Line Products
Change view
Click to view available options
Magnification Power:
10X, 30X
20X, 40X
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification Power
S81371 20X, 40X
S81372 10X, 30X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S81371 Supplier Motic™ Swift Line Supplier No. 453TBL10LED
Only null left
Add to Cart
Add to Cart

Motic Swift Line 450 Series Dual Magnification Intermediate Stereo Microscopes features rechargeable LED illumination and dual magnification with a rugged design and compact size.

  • Choice of 1X/3X or 2X/4X dual magnification 
  • Widefield 10X eyepieces with left side diopter adjustment
  • Interpupillary adjustment from 55 mm to 75 mm
  • Achromatic objectives are parfocalled and parcentered
  • 4-position light control: Top ony, bottom only, top and bottom, off
  • Top illumination: LED
  • Bottom illumination: LED
  • Rack and pinion focusing with slip-clutch and tension adjustment
  • Includes two 80 mm stage plates: Frosted glass and black/white contrast
  • Two locked-on stage clips

Specifications

Voltage 110/220 V
Magnification Power 20X, 40X
Objective Types Dual
Product Type Stereo Microscope
Model 453TBL-10-LED
Head Style Binocular
Lighting Type Dual LED
For Use With (Application) Intermediate Science Education
Eyepiece WF 10X
Quantity 1
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.