Learn More
Enterprise Technology Solutions Dual Monitor Mount Black
SDP
Catalog No. 503062509
Encompass_Preferred
Dual Monitor Mount For 27 Monitors 360 Degree Rotation 45 Degree/-45 Degree Tilt 90 Degree Pan Black Supports 20 Lb
Catalog No. 50-306-2509 Supplier Enterprise Technology Solutions Supplier No. MMMMM200B
Dual Monitor Mount For 27 Monitors 360 Degree Rotation 45 Degree/-45 Degree Tilt 90 Degree Pan Black Supports 20 Lb