Learn More
TA Instruments Dual MS FTIR Interface Kit for Q5000 IR, Connects Q5000 IR TGA to MS and FTIR, Works with Mass Spectrometer Interface Kit Without Library
Catalog No. 5000580635 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Dual MS/FTIR Interface Kit for the Q5000 IR is used in conjunction with the Q5000 IR Mass Spectrometer Interface Kit without library. It permits simultaneous connection of a Q5000 IR TGA to a MS and FTIR system.
Catalog No. 50-005-80635 Supplier TA Instruments Supplier No. 957574.901
Waters TA Instruments Dual MS/FTIR Interface Kit for the Q5000 IR is used in conjunction with the Q5000 IR Mass Spectrometer Interface Kit without library. It permits simultaneous connection of a Q5000 IR TGA to a MS and FTIR system.