Learn More
Trajan Scientific PEEK Tubing, Dual Layer Color Coded PEEK Tubing (Black) - 1/16" OD x .004" ID x 100FT, 6700 PSI, 460 BAR
Catalog No. 501385086
Encompass_Preferred
PEEK Tubing, Dual Layer Color Coded PEEK Tubing (Black) - 1/16" OD x .004" ID x 100FT, 6700 PSI, 460 BAR. Dual Layer PEEK tubing eliminates any concern that a critical sample stream could be contaminated by pigments used to color code the tubing. It looks like any other color-coded tubing at first glance, but a closer look reveals that the pigmented layer surrounds a separate but integrally-bonded inner layer of natural PEEK.
Catalog No. 50-138-5086 Supplier Trajan Scientific Supplier No. VAL.JRTD5804100
PEEK Tubing, Dual Layer Color Coded PEEK Tubing (Black) - 1/16" OD x .004" ID x 100FT, 6700 PSI, 460 BAR. Dual Layer PEEK tubing eliminates any concern that a critical sample stream could be contaminated by pigments used to color code the tubing. It looks like any other color-coded tubing at first glance, but a closer look reveals that the pigmented layer surrounds a separate but integrally-bonded inner layer of natural PEEK.