Learn More
Andwin Scientific Blue Film Heat-Seal Sterilization Pouch Convertors, 3x8
SDP
Catalog No. NC9771485
Encompass_Preferred
Blue Film Heat-Seal Sterilization Pouch Convertors For use in gas or steam sterilizers. High porosity for fast sterilization while maintaining effective bacterial barrier. May be heat-sealed or taped shut. Dual chemical indicators confirm sterilization ex
This item is not returnable. View return policy
Catalog No. NC9771485 Supplier Andwin Scientific Supplier No. 90308
Blue Film Heat-Seal Sterilization Pouch Convertors For use in gas or steam sterilizers. High porosity for fast sterilization while maintaining effective bacterial barrier. May be heat-sealed or taped shut. Dual chemical indicators confirm sterilization ex