Learn More
Andwin Scientific Gas/Steam Sterilization Tubing Convertors, Size: 5.1Wcmx30.5Lm., 1/RL
SDP
Catalog No. NC9747313
Encompass_Preferred
Gas/Steam Sterilization Tubing Convertors Blue film and surgical-grade paper combination includes two chemical process indicators. Cut tubing to the length needed insert item and heat-seal the open ends. Size: 5.1Wcmx30.5Lm., 1/RL
Catalog No. NC9747313 Supplier Andwin Scientific Supplier No. T90002
Gas/Steam Sterilization Tubing Convertors Blue film and surgical-grade paper combination includes two chemical process indicators. Cut tubing to the length needed insert item and heat-seal the open ends. Size: 5.1Wcmx30.5Lm., 1/RL