Learn More
Med Vet International Dual Pinch Nylon Clamp Large Up To 19mm, J0594A, 2/Pack
Catalog No. 5000583996
Encompass_Preferred
Larger size. Slides over tubing up to 3.4" or 19mmOD. 5/pk.
Catalog No. 50-005-83996 Supplier Med Vet International Supplier No. JOR594A
Larger size. Slides over tubing up to 3.4" or 19mmOD. 5/pk.