Learn More
Andwin Scientific DUAL POSITION PE CAP FOR 12 X
SDP
Catalog No. 50819821
Encompass_Preferred
The dual position cap is made from polyethylene. This product is specially designed to cover 12 x 75 mm tubes and protect its contents while permitting gas exchange. It can provide a secure sealed storage. This is sold in a pack of 1000 natural-colored caps.
Catalog No. 50-819-821 Supplier Andwin Scientific Supplier No. 14941030
The dual position cap is made from polyethylene. This product is specially designed to cover 12 x 75 mm tubes and protect its contents while permitting gas exchange. It can provide a secure sealed storage. This is sold in a pack of 1000 natural-colored caps.