Learn More
Euthanex Dual Preset Tank Regulator with Restrictors for both Mouse and Rat output.
Catalog No. NC1880333
Encompass
Dual Preset Regulator to distribute CO2"within AVMA recommended minimum fill rate for industry standard mouse and rat cages. Connects directly to the CO2 cylinder with CGA-320 fitting. (Optional CGA-940 Yoke fitting available. EP-2230 Hose Kit also available).
Catalog No. NC1880333 Supplier Euthanex Supplier No. EP2200
Dual Preset Regulator to distribute CO2"within AVMA recommended minimum fill rate for industry standard mouse and rat cages. Connects directly to the CO2 cylinder with CGA-320 fitting. (Optional CGA-940 Yoke fitting available. EP-2230 Hose Kit also available).