Learn More
Vetequip Inc DUAL PROCEDURE CIRCUIT COMPLET
SDP
Catalog No. NC9352805
Encompass_Preferred
Includes 2-liter inducation chamber, Y-circuit with stopcocks, 9mm, 12mm, and 14mm rodent nose cones, VaporGuards (2), and 6' 22mm evacuation tubing
Catalog No. NC9352805 Supplier Vetequip Inc Supplier No. 921400
Includes 2-liter inducation chamber, Y-circuit with stopcocks, 9mm, 12mm, and 14mm rodent nose cones, VaporGuards (2), and 6' 22mm evacuation tubing