Learn More
New Era Pump Systems Inc DUAL PUMP PLUMBING KIT
Catalog No. NC1416042
Encompass
For use with our continuous infusion system or dual infusion system. Provides dual check valves for two syringes, withdraw will pull fluid from a reservoir and infusion will output into a separate line. The kit is pre-assembled - just attach the syringes. Each dual check valve uses 6" of 1/8" ID tubing connected to a Y connector, one for reservoir and one for output; each Y-connector has 5' of tubing to make connections to your application. Two 50 mL NormJect Syringes are included.
Catalog No. NC1416042 Supplier New Era Pump Systems Inc Supplier No. PDKIT
For use with our continuous infusion system or dual infusion system. Provides dual check valves for two syringes, withdraw will pull fluid from a reservoir and infusion will output into a separate line. The kit is pre-assembled - just attach the syringes. Each dual check valve uses 6" of 1/8" ID tubing connected to a Y connector, one for reservoir and one for output; each Y-connector has 5' of tubing to make connections to your application. Two 50 mL NormJect Syringes are included.