Learn More
TA Instruments Dual Quartz Light Guide, Light Transmission Pathway, High Pressure Mercury Lamp to DSC Cell, 3 mm Diameter, 1000 mm Length
Catalog No. 5000580601 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Dual Quartz Light Guide provides a light transmission pathway from the high pressure mercury lamp to the DSC Cell. Each light guide has dimensions of 3 mm diameter by 1000 mm length.
Catalog No. 50-005-80601 Supplier TA Instruments Supplier No. 935012.002
Waters TA Instruments Dual Quartz Light Guide provides a light transmission pathway from the high pressure mercury lamp to the DSC Cell. Each light guide has dimensions of 3 mm diameter by 1000 mm length.