Learn More
Electron Microscopy Sciences Rotatrim Rolling Trimmer Dual-Rail Cutter, 30"
SDP
Catalog No. 5028392
Encompass_Preferred
For precision cutting through tough materials like cardboard, litho plates, mount board, and even 1/16" hardboard. Rotatrim also has a delicate touch, trimming film, paper, and dry mount tissue to hairline tolerances.
Known for its safe operation and best performance, the rotary blade is Sheffield tungsten steel, self-sharpening, and is completely shielded by a protective housing. Its self-sharpening blade assures consistent cutting quality.
Exclusively from Rotatrim, its double steel tube suspension guides the cutting head with the stability essential for accurate, repeatable cuts. An extruded aluminum squaring arm incorporates a combination inch/metric measuring scale and accepts the accessory side guide. A Transparent clamping strip, holds material in place without risk of damage.
Its durable, high pressure laminate finish keeps the baseboard warp-free, long lasting, and easy to clean.
Size: 30"
Catalog No. 50-283-92 Supplier Electron Microscopy Sciences Supplier No. 7488030
For precision cutting through tough materials like cardboard, litho plates, mount board, and even 1/16" hardboard. Rotatrim also has a delicate touch, trimming film, paper, and dry mount tissue to hairline tolerances.
Known for its safe operation and best performance, the rotary blade is Sheffield tungsten steel, self-sharpening, and is completely shielded by a protective housing. Its self-sharpening blade assures consistent cutting quality.
Exclusively from Rotatrim, its double steel tube suspension guides the cutting head with the stability essential for accurate, repeatable cuts. An extruded aluminum squaring arm incorporates a combination inch/metric measuring scale and accepts the accessory side guide. A Transparent clamping strip, holds material in place without risk of damage.
Its durable, high pressure laminate finish keeps the baseboard warp-free, long lasting, and easy to clean.
Size: 30"