Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Phosphate Test Kit
SDPSCI_ED

Catalog No. S72401 Shop All LaMotte Company Products
Change view
Click to view available options
Range:
Low: 1 to 10, High: 10 to 100 ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
S72401 Low: 1 to 10, High: 10 to 100 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S72401 Supplier LaMotte™ Supplier No. 440801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit tests for Phosphate using the Octa-Slide 2 Comparator.

  • Simple colorimeter
  • Simply react sampler per instructions and match color to the Octa-Slide viewer to determine orthophosphate concentration
  • Test System: Stannous Chloride, Octa-Slide 2 Comparator
  • Kit includes enough reagent to perform 50 tests
  • Range/Sensitivity:
  • Low: 1.0, 2.0, 3.0, 4.0, 5.0, 6.0, 8.0, 10.0 ppm PO43–
  • High: 10, 20, 30, 40, 50, 60, 80, 100 ppm PO43–

Specifications

For Use With (Application) Measures for industrial water boiling and cooling, laundry, sanitation, water and wastewater
Includes 2 x 30mL VM phosphate reagent, 30mL reducing reagent, plastic pipet dropping, 1mL plastic pipet, 0.5mL plastic pipet, 2 test tubes with caps, phosphate octa-slide 2 Bar, octa-slide 2 viewer
No. of Tests 100
Range Low: 1 to 10, High: 10 to 100 ppm
Test Method Phosphomolybdate complex/Stannous Chloride reduction
Type Phosphate Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.