Learn More
World Precision Instrument Dual Rate Syringe Pump
SDP
Catalog No. 502538574
Encompass
The intuitive SPLG-G88PLUS graphical user interface controlled with a large 7" LCD color touch screen display allows quick and easy setup. The display run screen presents the user with all key dispensing parameters in real time. Syringe tables containing all major syringe manufacturers allow simple selection of any compatible syringe size. Audible Alarms, Adjustable Force and Screen Lock are all features that are available with a touch of the screen.
Catalog No. 50-253-8574 Supplier World Precision Instrument Supplier No. SPLGG88PLUS
Add to Cart
Add to Cart
Request a Quote
The intuitive SPLG-G88PLUS graphical user interface controlled with a large 7" LCD color touch screen display allows quick and easy setup. The display run screen presents the user with all key dispensing parameters in real time. Syringe tables containing all major syringe manufacturers allow simple selection of any compatible syringe size. Audible Alarms, Adjustable Force and Screen Lock are all features that are available with a touch of the screen.