Learn More
USA Scientific Inc Dual rotor personal cent
Catalog No. 502745525
Encompass
Personal Minicentrifuge, 8 place 1.5/2.0 mL microcentrifuge tube rotor with adapters for both 0.5 mL and 0.2 mL tubes, and PCR strip rotor for up to 6,500 rpm 2,5000 x g
Catalog No. 50-274-5525 Supplier USA Scientific Inc Supplier No. 80305000
Personal Minicentrifuge, 8 place 1.5/2.0 mL microcentrifuge tube rotor with adapters for both 0.5 mL and 0.2 mL tubes, and PCR strip rotor for up to 6,500 rpm 2,5000 x g